Learn More
Biomatik Corporation Human Zinc Finger, BED Type Containing Protein 3 (ZBED3) CLIA Kit
Supplier:  Biomatik Corporation EKU09997
Encompass_Preferred
Human Zinc Finger, BED Type Containing Protein 3 (ZBED3) CLIA Kit for detecting ZBED3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q96IU2. Gene ID: 84327. Target Synonyms: Axin-interacting protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6238