Learn More
Biomatik Corporation Human Zinc Finger Homeobox Protein 4 (ZFHX4) ELISA Kit
Supplier:  Biomatik Corporation EKU08251
Encompass_Preferred
Human Zinc Finger Homeobox Protein 4 (ZFHX4) ELISA Kit for detecting ZFHX4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q86UP3. Gene ID: 79776. Target Synonyms: ZFH4; Zinc finger homeodomain protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8686