Learn More
Biomatik Corporation Human Zinc Finger Protein 10 (ZNF10) ELISA Kit
Supplier:  Biomatik Corporation EKU12415
Encompass_Preferred
Human Zinc Finger Protein 10 (ZNF10) ELISA Kit for detecting ZNF10 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21506. Gene ID: 7556. Target Synonyms: KOX1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7968