Learn More
Biomatik Corporation Human Zinc Ribbon Domain Containing Protein 1 (ZNRD1) ELISA Kit
Supplier:  Biomatik Corporation EKU08254
Encompass_Preferred
Human Zinc Ribbon Domain Containing Protein 1 (ZNRD1) ELISA Kit for detecting ZNRD1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.107ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9P1U0. Gene ID: 30834. Target Synonyms: HTEX-6; Rpa12; hZR14; Tctex-6; DNA-directed RNA polymerase I subunit RPA12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7547