Learn More
American Research Products Inc Human Zinc transporter 8 SLC3
Catalog No. 5000328179
Encompass
Human Zinc transporter 8 (SLC30A8) ELISA kit from CUSABIO Detection Range 87 5 pg/ml-1400 pg/ml Sensitivity 17 5pg/ml Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28179 Supplier American Research Products Inc Supplier No. CSBEL021573HU
Human Zinc transporter 8 (SLC30A8) ELISA kit from CUSABIO Detection Range 87 5 pg/ml-1400 pg/ml Sensitivity 17 5pg/ml Sample serum plasma tissue homogenates cell lysates Method Sandwich