Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ZMIZ2 (aa 712-827) Control Fragment Recombinant Protein

Catalog No. RP89718
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP89718 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP89718 Supplier Invitrogen™ Supplier No. RP89718
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59263. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ZIMP7, also known as ZMIZ2, is a novel PIAS (protein inhibitor of activated signal transducer and activator of transcription)-like protein and a transcriptional coactivator. ZIMP7 is expressed most abundantly in testis. The C-terminal proline-rich domain possesses a significant intrinsic transcriptional activity and this activity is inhibited by the N-terminus in the full-length ZIMP7. ZIMP7 and the related protein ZIMP10 interact with PIAS3 and enhances Androgen Receptor (AR)- mediated transcription. The interaction between ZIMP7 and SWI/SNF complex suggests a possible role for ZIMP7 in chromatin modification.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NF64
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 83637
Name Human ZMIZ2 (aa 712-827) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias D11Bwg0280e; HRIHFB2007; hZIMP7; KIAA1886; NET27; PIAS-like protein Zimp7; TRAFIP20; Zimp7; zinc finger MIZ domain-containing protein 2; zinc finger MIZ-type containing 2; zinc finger, MIZ-type containing 2; Zmiz2
Common Name ZMIZ2
Gene Symbol ZMIZ2
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LMPSVMEMIAALGPGAAPFAPLQPPSVPAPSDYPGQGSSFLGPGTFPESFPPTTPSTPTLAEFTPGPPPISYQSDIPSSLLTSEKSTACLPSQMAPAGHLDPTHNPGTPGLHTSNL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.