Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ZNF326 (aa 398-483) Control Fragment Recombinant Protein

Catalog No. RP94401
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP94401 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP94401 Supplier Invitrogen™ Supplier No. RP94401
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55867. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Core component of the DBIRD complex, a multiprotein complex that acts at the interface between core mRNP particles and RNA polymerase II (RNAPII) and integrates transcript elongation with the regulation of alternative splicing: the DBIRD complex affects local transcript elongation rates and alternative splicing of a large set of exons embedded in (A + T)-rich DNA regions. May play a role in neuronal differentiation and is able to bind DNA and activate expression in vitro. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5BKZ1
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 284695
Name Human ZNF326 (aa 398-483) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 5730470H14Rik; DBIRD complex subunit ZNF326; dJ871E2.1; dJ871E2.1 (novel protein (ortholog of mouse zinc finger protein Zan75)); Zan75; Zfp326; zinc finger protein 326; zinc finger protein 326; DBIRD complex subunit ZNF326; zinc finger protein interacting with mRNPs; Zinc finger protein interacting with mRNPs and DBC1; zinc finger protein-associated with nuclear matrix of 75 kDa; Zird; ZNF326; ZNF-protein interacting with nuclear mRNPs and DBC1
Common Name ZNF326
Gene Symbol ZNF326
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence HMMKVETVHCSACSVYIPALHSSVQQHLKSPDHIKGKQAYKEQIKRESVLTATSILNNPIVKARYERFVKGENPFEIQDHSQDQQI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.