Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ZNF350 (aa 58-182) Control Fragment Recombinant Protein

Catalog No. RP99731
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP99731 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP99731 Supplier Invitrogen™ Supplier No. RP99731
  • $283.00
    $212.25 / Each of 1

    Save $70.75

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (25%), Rat (25%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-81951. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ZBRK1 is a member of the Kruppel-associated box-zinc finger protein (KRAB-ZFP) protein family of transcriptional repressors. ZBRK1 has been shown to not only be up-regulated in senescent fibroblasts but also interacts with BRCA1, a transcriptional regulator with tumor suppressor activity. ZBRK1 binds to a specific sequence within the GADD45 gene, supporting the assembly of a nuclear complex containing BRCA1 and repressing its transcription. Recent experiments demonstrate that ZBRK1 can homo-oligomerize in vitro and in vivo through a C-terminal transcriptional repression domain (CTRD), and this oligomerization facilitates the ZBRK1-directed transcriptional repression through ZBRK1 response elements. These results suggest that higher order ZBRK1 oligomers may assemble through both protein-DNA and CTRD-dependent protein-protein interactions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9GZX5
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 59348
Name Human ZNF350 (aa 58-182) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias KRAB zinc finger protein ZFQR; ZBRK1; ZFQR; Zinc finger and BRCA1-interacting protein with a KRAB domain 1; zinc finger protein 350; Zinc finger protein ZBRK1; zinc-finger protein ZBRK1; ZNF350
Common Name ZNF350
Gene Symbol ZNF350
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FKLEQGEQLWTIEDGIHSGACSDIWKVDHVLERLQSESLVNRRKPCHEHDAFENIVHCSKSQFLLGQNHDIFDLRGKSLKSNLTLVNQSKGYEIKNSVEFTGNGDSFLHANHERLHTAIKFPASQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.