Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ZNF536 Synthetic Peptide

Catalog No. PEP0968
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0968 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0968 Supplier Invitrogen™ Supplier No. PEP0968
Add to Cart
Add to Cart

Synthetic Peptide

ZNF536 Synthetic Peptide, PEP-0968, from Invitrogen.

ZNF536 is a recently identified zinc-finger protein that is expressed primarily in the developing nervous system and the cerebral cortex, hippocampus, and hypothalamus. ZNF536 possess ten zinc fingers and interacts with CtBP1, a corepressor for gene transcription. It is most closely related to transcriptional repressor ZNF219. Overexpression of ZNF536 in embryonic stem cells dramatically reduced the mRNA levels of neuronal marker genes such as Pax6, MAP2, and beta-tubulin III following retinoic acid (RA)-induced differentiation, while depletion of ZNF536 via RNAi resulted in elevated mRNA levels of these genes, indicating its role in inhibiting neuronal cell differentiation. Overexpression of RA receptor a rescues the inhibitory role of ZNF536, suggesting that ZNF536 might inhibit RA response element-mediated transcriptional activity.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 9745
Species Human
Purification Method purified
Accession Number O15090
Common Name Human ZNF536
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol ZNF536
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.