Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ZNF667 Synthetic Peptide

Catalog No. PEP0883 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0883 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0883 Supplier Invitrogen™ Supplier No. PEP0883
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 17 amino acids near the amino terminus of human MIPU1. PEP-0883 can be used as a blocking peptide with polyclonal antibody PA5-20769.

Mipu1, also known as zinc finger protein 667 or ZNF667, encodes a nuclear-localized protein containing 14 carboxy-terminal zinc finger motifs and an amino-terminal KRAB domain. This protein is highly expressed in heart and brain and is upregulated in rat heart after a transient ischemia-reperfusion procedure. Overexpression experiments suggest that Mipu1 suppresses the transcriptional activities of AP-1 and SRE in the MAPK signaling pathway and thus may play a role in the pathogenesis of cardiac and vascular disease. At least four isoforms of MIPU1 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 63934
Species Human
Purification Method purified
Accession Number Q5HYK9
Common Name ZNF667
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias A830025F02Rik; Mip1; MIPU1; myocardial ischemic preconditioning upregulated 1; myocardial ischemic preconditioning upregulated 1 ortholog; Myocardial ischemic preconditioning up-regulated protein 1; Zfp667; Zinc finger protein 667; Znf667
Gene Symbol ZNF667
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.