Learn More
ACROBiosystems Human ZNRF3 protein, His Tag
Supplier:  ACROBiosystems ZN3H52H31MG
Encompass_Preferred
Recombinant Protein;1MG;Human ZNRF3, His Tag (ZN3-H52H3) is expressed from human 293 cells (HEK293). It contains AA Lys 56 - Met 219 (Accession # Q9ULT6-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.0 kDa.
Catalog No. 50-205-4892