Learn More
Abcam Native Human IgG FC fragment protein
Supplier:  Abcam AB902851MG
Encompass_Preferred
Native Human IgG FC fragment protein is a Full Length protein with >95% purity and suitable for SDS-PAGE. The predicted molecular weight of ab90285 native protein is 50 kDa. - Save time and ensure accurate results - use our IgG Fc fragment protein as a control - Cited in over 10 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0369482