Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Humanized mAb Mass Check Standard, Reversed Phase, Solid, 80 μg

Catalog No. 502038913 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The Humanized mAb Mass Check Standard supports the analysis of intact proteins by LC-MS, a preferred approach for characterizing biopharmaceuticals. It enables acquiring intact mass information, quickly confirming sequence and post-translational modifications, and interrogating product-related impurities.

  • Supports benchmarking, proficiency testing, and system suitability for intact protein analysis.
  • Stabilized and lyophilized format of NIST Reference Material 8671.
  • Each vial contains approximately 80 μg of the standard.
  • Manufactured to exact specifications and completely traceable.
  • Stable for seven days at 4–8 °C after reconstitution.
  • Separation mode: Reversed Phase.
  • Formulation: Solid.
  • Storage condition: −20°C ± 10°C (Freezer).
  • Shelf life: 36 Months.
  • Shipping condition: Ambient.
  • Application: Protein.

Catalog No. 50-203-8913 Supplier Waters Corp Supplier No. 186009125
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Humanized mAb Mass Check Standard supports the analysis of intact proteins by LC-MS, a preferred approach for characterizing biopharmaceuticals. It enables acquiring intact mass information, quickly confirming sequence and post-translational modifications, and interrogating product-related impurities.

  • Supports benchmarking, proficiency testing, and system suitability for intact protein analysis.
  • Stabilized and lyophilized format of NIST Reference Material 8671.
  • Each vial contains approximately 80 μg of the standard.
  • Manufactured to exact specifications and completely traceable.
  • Stable for seven days at 4–8 °C after reconstitution.
  • Separation mode: Reversed Phase.
  • Formulation: Solid.
  • Storage condition: −20°C ± 10°C (Freezer).
  • Shelf life: 36 Months.
  • Shipping condition: Ambient.
  • Application: Protein.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.