Learn More
Abcam Human/Mouse LRRK2 Antibody Pair - BSA and Azide free, 10 x 96 Tests
Catalog No. 502689763
Encompass_Preferred
Human/Mouse LRRK2 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human/Mouse LRRK2.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-9763 Supplier Abcam Supplier No. AB30774710X96TESTS
Human/Mouse LRRK2 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human/Mouse LRRK2.
The product is subject to the following: Abcam Restricted Use Statement