Learn More
Biomatik Corporation Human/Mouse/Rat Cytochrome C (CYCS) ELISA Kit
Supplier:  Biomatik Corporation EKU08539
Encompass_Preferred
Human/Mouse/Rat Cytochrome C (CYCS) ELISA Kit for detecting CYCS in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P62897. Gene ID: 13063. Target Synonyms: Cyt-C; HCS; CYC; CytC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8656