Learn More
Biomatik Corporation Human/Mouse/Rat Mothers Against Decapentaplegic Homolog 1 (Smad1) ELISA Kit
Supplier:  Biomatik Corporation EKU09013
Encompass_Preferred
Human/Mouse/Rat Mothers Against Decapentaplegic Homolog 1 (Smad1) ELISA Kit for detecting Smad1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.132ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P70340; P97588. Gene ID: 17125; 25671. Target Synonyms: BSP1; JV4-1; JV41; MADH1; MADR1; SMAD Family Member 1; Mad-related protein 1; Transforming growth factor-beta-signaling protein 1; Mothers against DPP homolog 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4634