Learn More
Biomatik Corporation Human/Mouse/Rat Signal Transducer And Activator Of Transcription 5B (STAT5B) ELISA Kit
Supplier:  Biomatik Corporation EKU09075
Encompass_Preferred
Human/Mouse/Rat Signal Transducer And Activator Of Transcription 5B (STAT5B) ELISA Kit for detecting STAT5B in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P51692. Gene ID: 6777. Target Synonyms: STAT5-B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6125