Learn More
Biomatik Corporation Human/Mouse/Rat Signal Transducer And Activator Of Transcription 4 (STAT4) ELISA Kit
Supplier:  Biomatik Corporation EKU09077
Encompass_Preferred
Human/Mouse/Rat Signal Transducer And Activator Of Transcription 4 (STAT4) ELISA Kit for detecting STAT4 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14765. Gene ID: 6775. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8570