Learn More
Biomatik Corporation Human/Mouse/Rat Stathmin 1 (STMN1) ELISA Kit
Supplier:  Biomatik Corporation EKU09139
Encompass_Preferred
Human/Mouse/Rat Stathmin 1 (STMN1) ELISA Kit for detecting STMN1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P16949. Gene ID: 3925. Target Synonyms: LAP18; C1orf215; Lag; OP18; PP17; PP19; PR22; SMN; Metablastin; Prosolin; Oncoprotein 18; Leukemia-associated phosphoprotein p18. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5092