Learn More
MyBioSource SCD14 ST ELISA Kit
SDP
Supplier:  MyBioSource MBS380091696STRIPWELLS
Encompass_Preferred
The MyBioSource SCD14-ST ELISA Kit quantitatively measures human SCD14 in various biological fluids using a sandwich enzyme immunoassay. It involves a monoclonal antibody-coated 96-well plate, biotin-conjugated polyclonal antibody, and HRP. Color change upon adding TMB substrate and Stop Solution is measured at 450 nm to determine SCD14 levels.
Catalog No. NC2601118