Learn More
Eurofins Discoverx huMAP Kinase 2/Erk2, active, 250 µg. (14-550M)
Supplier:  Eurofins Discoverx 14550M
Encompass
N-terminal GST-tagged recombinant, human full length MAP Kinase 2/Erk2, Activated with MEK1 (14-429). For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8954