Learn More
Humboldt Ball Retaining Rings for Softening Point Apparatus
SDP
Catalog No. 015725
Change view
Click to view available options
For Use With (Equipment):
Softening point apparatus
Catalog No. For Use With (Equipment)
01-572-5 Softening point apparatus
1 options
Catalog No. 01-572-5 Supplier Humboldt Supplier No. H1588
Add to Cart
Add to Cart
Fit over 01-571 ring for centering steel ball on sample.
Specifications
Product Type Ball Retaining Ring
For Use With (Equipment) Softening point apparatus