Learn More
Humboldt Brass Shouldered Rings for Softening Point Apparatus
SDP
Catalog No. 01571
Change view
Click to view available options
For Use With (Equipment):
Softening point apparatus
Catalog No. For Use With (Equipment)
01-571 Softening point apparatus
1 options
Catalog No. 01-571 Supplier Humboldt Supplier No. H1575
Specifications
Product Type Brass Shouldered Ring
Dimensions (L x W x H) 23 x 23 x 2 mm
For Use With (Equipment) Softening point apparatus
Height (Metric) 2 mm