Learn More
Humboldt Sharpener for Cork Borers
SDP
Catalog No. 07865
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
07-865 Each
1 options
Catalog No. 07-865 Supplier Humboldt Supplier No. H9680
Add to Cart
Add to Cart
Consists of a brass cone and heavy steel-cutting edge mounted on a wooden handle.
Simply slide borer over brass cone (boring edge down), sharpen by grinding edge against hard tool steel blade.
Specifications
Product Type Cork Borer Sharpener
For Use With (Application) To sharpen borers from 0.18 to 0.93 in.
For Use With (Equipment) For cork borers
Includes Brass cone and heavy Steel-cutting edge mounted in a wooden handle
Quantity Each