Learn More
Humboldt Softening Point Apparatus
SDP
Catalog No. 01551
Change view
Click to view available options
For Use With (Application):
Brass Ring and Ball Method (Double)
Catalog No. For Use With (Application)
01-551 Brass Ring and Ball Method (Double)
1 options
Catalog No. 01-551 Supplier Humboldt Supplier No. H1569
Add to Cart
Add to Cart
Apparatus, Softening Point; Humboldt; With beaker; Shouldered ring-and-ball; Consists of beaker with shelving, two std. balls, two rings, cover; Samples held within horizontal rings; Steel balls placed on top of samples
In accordance with ASTM D 36, E 28, and D 2398 tests
Shouldered ring-and-ball apparatus for softening point determinations according to ASTM D 36, E 28, and D 2398. Apparatus consists of beaker with shelving, balls, rings, cover. Samples are held within horizontal rings. Steel balls are placed on top of samples. As sample, which is submerged in water, is heated, weight of ball forces softened sample downward. Softening point is temperature at which sample falls 1 in. (2.5cm) to bottom shelf. Apparatus comes with two standard balls, two 01-571 shouldered rings, 02-540N beaker, two shelves (1 in. [2.5cm] apart) with support rods, metal cover and 01-572-5 ball retaining rings.
Order Info
(Order thermometers separately.)
Specifications
Product Type Softening Point Apparatus
Certifications/Compliance Meet ASTM D36, E28, AASHTO T53
For Use With (Application) Brass Ring and Ball Method (Double)
Includes 800 mL beaker, 2 standard balls, 2 shouldered rings and 2 ball centering guides