Learn More
Caron Products And Services HUMD304 EARTH-FRIENDLY GVAPOR
Catalog No. NC1976382
Encompass_Preferred
Earth-friendly gVapor(steam-free humidity) additive control from ambient to 95% RH with display/readout for Caron Model: 7400-25 & 7400-33 series CO2 incubators. Factory installed at the time of order.
Catalog No. NC1976382 Supplier Caron Products And Services Supplier No. HUMD304
Earth-friendly gVapor(steam-free humidity) additive control from ambient to 95% RH with display/readout for Caron Model: 7400-25 & 7400-33 series CO2 incubators. Factory installed at the time of order.