Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ HuMEC Supplement Kit

Catalog No. 12755013
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-755-013 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-755-013 Supplier Gibco™ Supplier No. 12755013
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HuMEC Supplement Kit

HuMEC Supplement Kit contains growth factors and other components and is designed to work with HuMEC Basal Serum-Free Medium (12753018). Together, these products are engineered specifically for the culture of human mammary epithelial cells (A10565). This translates to greater experimental validity, more reproducible results, and reduction of lot-to-lot variability and aberrations that can occur with the addition of serum. Human mammary epithelial cells cultured in HuMEC Medium have gene expression profiles consistent with normal mammary epithelial cells, as tested on a cDNA microarray.

Applications:
HuMEC Medium is optimized specifically for the growth of human mammary epithelial cells, and has been performance tested in a growth assay using normal human mammary epithelial cells. HuMEC Medium promotes superior growth of normal human mammary epithelial cells, making it ideal for researchers in breast cancer, drug discovery, and other aspects of breast biology.

Contents and Storage:
The HuMEC Supplement Kit includes 5 ml of a supplement mix containing epidermal growth factor, hydrocortisone, isoproterenol, transferrin, and insulin, and 25 mg of bovine pituitary extract. Store the supplement mix at -5°C to -20°C.

Related Links:

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Epithelial Cells (Mammary)
Culture Type Mammalian Cell Culture
Product Type HuMEC Supplement
Serum Level Serum-Free
Sterility Sterile-filtered
Quantity 1 Kit
Species Human

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.