Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Humic acid sodium salt, tech. 50-60% (as humic acid)

Catalog No. p-7051675
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4174822 100 g
AA4174836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4174822 Supplier Thermo Scientific Chemicals Supplier No. 04174822
Only null left
Add to Cart

CAS: 68131-04-4 | C9H8Na2O4 | 226.139 g/mol

Humic acid sodium salt is a principal component of humic substances. It boosts plant growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68131-04-4
Molecular Formula C9H8Na2O4
Molecular Weight (g/mol) 226.139
MDL Number MFCD00135560
InChI Key HMGUIQPKFUZDPV-UHFFFAOYSA-L
Synonym humic acid sodium salt, sodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, sodium humate, disodium bicyclo 2.2.1 hept-5-en-2,3-dicarboxylate, disodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, bicyclo 2.2.1 hepta-5-ene-2,3-dicarboxylic acid disodium salt
PubChem CID 21908873
IUPAC Name disodium;bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate
SMILES C1C2C=CC1C(C2C(=O)[O-])C(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >300°C
Color Black
Merck Index 14,4752
Solubility Information Soluble in water (33.3 mg in 1 mL)
Percent Purity 50 to 60%
Grade Technical
Quantity 100 g
Physical Form Powder
Assay (as humic acid)
Chemical Name or Material Humic acid sodium salt
Hazard Category H315-H319-H335-H500
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 268-608-0
RTECSNumber MT6550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.