Learn More
Med Vet International Humidification Management Exchanger, over 75 lbs, J0606B, 5 Pack
Catalog No. 5000581319
Encompass_Preferred
In human anesthesia the prevention of moisture andheat loss is a critical consideration. The smaller patientis especially a concern because hypothermia is more likely to occur. Humidification management exchangers are routinely used in human anesthesia. Veterinarians are starting to recognize the value of these products andmake them part of their anesthetic regime.The Humid-Vent is a compact connector that is a heat and moisture exchanger. They are placed in the breathing circuit between the "Y" piece (or "T" piece for non-rebreathing) and the endotracheal tube. Inside the connector is a special medium consisting of hygroscopic, naturally absorbent microwell paper. The Humid-Vent can be used up to 24 hours. After 24 hours they lose their effectiveness and must be replaced. In veterinary use they are typically replaced weekly. 5/box.
Catalog No. 50-005-81319 Supplier Med Vet International Supplier No. JOR606B
In human anesthesia the prevention of moisture andheat loss is a critical consideration. The smaller patientis especially a concern because hypothermia is more likely to occur. Humidification management exchangers are routinely used in human anesthesia. Veterinarians are starting to recognize the value of these products andmake them part of their anesthetic regime.The Humid-Vent is a compact connector that is a heat and moisture exchanger. They are placed in the breathing circuit between the "Y" piece (or "T" piece for non-rebreathing) and the endotracheal tube. Inside the connector is a special medium consisting of hygroscopic, naturally absorbent microwell paper. The Humid-Vent can be used up to 24 hours. After 24 hours they lose their effectiveness and must be replaced. In veterinary use they are typically replaced weekly. 5/box.