Learn More
GENIE GROUP Humidity Alert II
Catalog No. NC0979706
Encompass_Preferred
This item has a minimum qty of 3 per supplier requirements.
Designed to monitor relative humidity, & temp. Features include a super large display, C/F switchable, Min/Max w/ reset function, low battery indication & a built-in stand for sitting on a desk & hardware for mounting on a wall. Designed to monitor relative humidity, temp & dew point. The meter warns users of environmental conditions that are out of a desired range - ideal for preventing mold growth. The humidity/temp probe is connected to an 18 inch long cable & can be clipped onto the meter or used in a remote location. What's Included: mounting bracket & one AAA battery. 1 inch Digits on super large LCD Max/Min w/ ''reset'' function RH accuracy: 5%RH temp accuracy: 1.8F, (1C) temp range: 14 to 140F (-10 to 60C) Humidity range: 10 to 99% RH Rear calibration adjustment pot Probe clips onto meter or extends on 18" (457mm) cable Probe clips onto meter or extends on 18" (457mm) cable
Catalog No. NC0979706 Supplier GENIE GROUP Supplier No. 445815
This item has a minimum qty of 3 per supplier requirements.
Designed to monitor relative humidity, & temp. Features include a super large display, C/F switchable, Min/Max w/ reset function, low battery indication & a built-in stand for sitting on a desk & hardware for mounting on a wall. Designed to monitor relative humidity, temp & dew point. The meter warns users of environmental conditions that are out of a desired range - ideal for preventing mold growth. The humidity/temp probe is connected to an 18 inch long cable & can be clipped onto the meter or used in a remote location. What's Included: mounting bracket & one AAA battery. 1 inch Digits on super large LCD Max/Min w/ ''reset'' function RH accuracy: 5%RH temp accuracy: 1.8F, (1C) temp range: 14 to 140F (-10 to 60C) Humidity range: 10 to 99% RH Rear calibration adjustment pot Probe clips onto meter or extends on 18" (457mm) cable Probe clips onto meter or extends on 18" (457mm) cable