Learn More
Grainger REED Instruments Humidity Meter Calibration Kit, 33% and 75% RH, 2-Piece Set
Catalog No. 502761679
Encompass_Preferred
This humidity calibration kit is designed to verify the accuracy of compatible humidity meters and probes. The kit facilitates precise calibration by providing 33% and 75% relative humidity standards.
  • Includes R9933 (33% humidity calibration standard) and R9975 (75% humidity calibration standard)
  • Ensures ±1% relative humidity accuracy at 77°F
  • Compatible with probes up to 15 mm (0.6 in.) in diameter
  • Designed for use with 8706, R6001, R6200, R9900, and SD-3007 models
Catalog No. 50-276-1679 Supplier Grainger Supplier No. 161C49
This humidity calibration kit is designed to verify the accuracy of compatible humidity meters and probes. The kit facilitates precise calibration by providing 33% and 75% relative humidity standards.
  • Includes R9933 (33% humidity calibration standard) and R9975 (75% humidity calibration standard)
  • Ensures ±1% relative humidity accuracy at 77°F
  • Compatible with probes up to 15 mm (0.6 in.) in diameter
  • Designed for use with 8706, R6001, R6200, R9900, and SD-3007 models