Learn More
Mitegen Humidity Control Workstation
SDP
Catalog No. 5000014202
Encompass_Preferred
The MiTeGen Watershed is a humidity-controlled workstation designed to address drop dehydration during crystal harvesting particularly in protein crystallography This innovative device provides a stable regulated environment for sensitive sample preparation maximizing high-quality reproducible diffraction data By maintaining sample hydration the Watershed helps researchers optimize crystal harvesting and refine treatment protocols improving efficiency and results Key features include precise humidity control to prevent dehydration and simplified crystal mounting with ergonomic design enhancing workflow The Watershed is versatile for biomolecular crystallography cell biology and assisted reproduction offering a convenient solution to sample dehydration in labs of all sizes
Catalog No. 50-000-14202 Supplier Mitegen Supplier No. MWS1803201
Add to Cart
Add to Cart
Request a Quote
The MiTeGen Watershed is a humidity-controlled workstation designed to address drop dehydration during crystal harvesting particularly in protein crystallography This innovative device provides a stable regulated environment for sensitive sample preparation maximizing high-quality reproducible diffraction data By maintaining sample hydration the Watershed helps researchers optimize crystal harvesting and refine treatment protocols improving efficiency and results Key features include precise humidity control to prevent dehydration and simplified crystal mounting with ergonomic design enhancing workflow The Watershed is versatile for biomolecular crystallography cell biology and assisted reproduction offering a convenient solution to sample dehydration in labs of all sizes