Learn More
Perkin Elmer US LLC Humidity Generator Sensor Assembly - Calibrated
Catalog No. 502113475
Encompass_Preferred
The Humidity Generator regulates the relative humidity of clean, ambient air for routine humidity experimentation. The accessory flows ambient air through an integrated desiccant reservoir to clean the air. Next, it diverts a portion of this air through an 80 mL heated water reservoir and then through a heated connection tube to the DMA analyzer. This generator is compatible with the DMA 8000 analyzer.
Catalog No. 50-211-3475 Supplier Perkin Elmer US LLC Supplier No. N5330402
The Humidity Generator regulates the relative humidity of clean, ambient air for routine humidity experimentation. The accessory flows ambient air through an integrated desiccant reservoir to clean the air. Next, it diverts a portion of this air through an 80 mL heated water reservoir and then through a heated connection tube to the DMA analyzer. This generator is compatible with the DMA 8000 analyzer.