Learn More
BRINSEA PRODUCTS Ova-Easy Advance Humidity Pump Silicone Tubing
Catalog No. NC2184515
Encompass_Preferred
Smaller bore silicone tubing used for interconnecting the incubator or hatcher to the water reservoir of the Ova-Easy Advance Humidity Pump (part # NC1728213) which provides fully automatic control of humidity when paired with any of the Brinsea Ova-Easy Cabinet Incubators. Larger water tubing (part # NC2184510) is used around the pump capstan. For optimal performance, the tubing will need replacing about every 3 months depending on usage. Sold by the foot.
This item is not returnable. View return policy
Catalog No. NC2184515 Supplier BRINSEA PRODUCTS Supplier No. 14.11
Smaller bore silicone tubing used for interconnecting the incubator or hatcher to the water reservoir of the Ova-Easy Advance Humidity Pump (part # NC1728213) which provides fully automatic control of humidity when paired with any of the Brinsea Ova-Easy Cabinet Incubators. Larger water tubing (part # NC2184510) is used around the pump capstan. For optimal performance, the tubing will need replacing about every 3 months depending on usage. Sold by the foot.