Learn More
Research Products International Corp Humidity Sponges, Single Use, 40 per Case
SDP
Catalog No. 50999140
Encompass
Humidity Sponges are ideal for providing dry conditions in laboratory applications. Humidity Sponges have a high moisture absorbency of up to 37% and one bag will maintain dry conditions inside a container with a two gallon volume.
The non-toxic desiccant is enclosed within a breathable polyethylene bag. Indicating material changes color to signal when full moisture capacity has been reached.
Catalog No. 50-999-140 Supplier Research Products International Corp Supplier No. 283150
Humidity Sponges are ideal for providing dry conditions in laboratory applications. Humidity Sponges have a high moisture absorbency of up to 37% and one bag will maintain dry conditions inside a container with a two gallon volume.
The non-toxic desiccant is enclosed within a breathable polyethylene bag. Indicating material changes color to signal when full moisture capacity has been reached.