Learn More
Rotronic™ Humidity standards
SDP
Catalog No. 116507 Shop All Rotronic Instrument Corp Products
Click to view available options
Type:
Humidity Standard
Humidity Standard (Certified)
Solution Type:
0.5% RH
10% RH
35% RH
80% RH
95% RH
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Solution Type
11-650-7 Humidity Standard 95% RH
11-650-12 Humidity Standard (Certified) 0.5% RH
11-650-13 Humidity Standard (Certified) 10% RH
11-650-4 Humidity Standard 10% RH
11-650-5 Humidity Standard 35% RH
11-650-6 Humidity Standard 80% RH
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 11-650-7 Supplier Rotronic™ Supplier No. EA95
Includes
SCS traceable calibration certificate, textile pads, Calibration instructions
For on-site calibration and adjustment of ROTRONIC probes with handheld instrument
With the humidity standards, a calibration device and HW4 software, running on a PC this is easily achieved.
  • Ampoules contain unsaturated salt solutions
  • Simple and safe on-site calibration method
  • Unlimited storage life
Specifications
Packaging 0.95
Certificate of Analysis SCS traceable calibration certificate, textile pad, calibration instruction
Technique On-site Calibration and Adjustment of Rotronic Probe
For Use With (Equipment) Rotronic Instrument
Solution Type 95% RH
Type Humidity Standard