Learn More
Cole-Parmer Extech 44550 Humidity/Temperature Pen, 20 to 90% RH/, 14 to 122F
Catalog No. 50993034 Shop All Cole-Parmer Products
Encompass
Pen displays relative humidity and temperature simultaneously. Min/max function displays and holds the maximum humidity and temperature values since power-up in memory; simply press the clear button to reset. Featuring an on/off button, and selectable °F/°C temperature.
Catalog No. 50-993-034 Supplier Cole-Parmer Supplier No. 3780314
Pen displays relative humidity and temperature simultaneously. Min/max function displays and holds the maximum humidity and temperature values since power-up in memory; simply press the clear button to reset. Featuring an on/off button, and selectable °F/°C temperature.