Learn More
Cayman Chemical Human Activator Protein-1 Reporter Assay System, 96 Well
SDP
Catalog No. 502919440
Encompass_Preferred
Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryopreservation method yields high cell viability post-thaw and provides the convenience of immediately dispensing healthy reporter cells into assay plates. There is no need for intermediate preparatory steps such as the spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup.
Catalog No. 50-291-9440 Supplier Cayman Chemical Supplier No. 3722896 WELL
Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryopreservation method yields high cell viability post-thaw and provides the convenience of immediately dispensing healthy reporter cells into assay plates. There is no need for intermediate preparatory steps such as the spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup.