Learn More
Cayman Chemical Humn Adrenoceptr Beta 1 Rep 96
SDP
Catalog No. 502944857
Encompass_Preferred
INDIGO's assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to ADRB1 Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator Norepinephrine, Luciferase Detection Reagents, and a cell culture-ready assay plate.
Catalog No. 50-294-4857 Supplier Cayman Chemical Supplier No. 4089396 WELL
INDIGO's assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to ADRB1 Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator Norepinephrine, Luciferase Detection Reagents, and a cell culture-ready assay plate.