Learn More
Cayman Chemical Humn Adrenoceptr Beta 2 Rep
SDP
Catalog No. 502944690
Encompass_Preferred
INDIGO's assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to ADRB2 Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator Isoproterenol, Luciferase Detection Reagents, and a cell culture-ready assay plate.
Catalog No. 50-294-4690 Supplier Cayman Chemical Supplier No. 40894384 WELL
INDIGO's assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to ADRB2 Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator Isoproterenol, Luciferase Detection Reagents, and a cell culture-ready assay plate.