Learn More
Cayman Chemical Humn Adrenoceptr Beta 3 Rep 96
SDP
Catalog No. 502945361
Encompass_Preferred
INDIGO's assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to ADRB3 Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator Isoproterenol, Luciferase Detection Reagents, and a cell culture-ready assay plate.
Catalog No. 50-294-5361 Supplier Cayman Chemical Supplier No. 4263296 WELL
INDIGO's assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to ADRB3 Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator Isoproterenol, Luciferase Detection Reagents, and a cell culture-ready assay plate.