Learn More
Cayman Chemical Human Arginine Vasopressin Receptor 1B Reporter Assay System, 384 Well
SDP
Catalog No. 502945273
Encompass_Preferred
INDIGO's assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to AVPR1B Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator Vasopressin, Luciferase Detection Reagents, and a cell culture-ready assay plate.
Catalog No. 50-294-5273 Supplier Cayman Chemical Supplier No. 42633384 WELL
INDIGO's assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to AVPR1B Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator Vasopressin, Luciferase Detection Reagents, and a cell culture-ready assay plate.