Learn More
Cayman Chemical Human Arginine Vasopressin Receptor 2 Reporter Assay System, 384 Well
SDP
Catalog No. 502945239
Encompass_Preferred
This assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to AVPR2 Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator vasopressin, Luciferase Detection Reagents, a cell culture-ready assay plate, and a detailed assay protocol.
Catalog No. 50-294-5239 Supplier Cayman Chemical Supplier No. 42634384 WELL
This assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to AVPR2 Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator vasopressin, Luciferase Detection Reagents, a cell culture-ready assay plate, and a detailed assay protocol.