Learn More
Cayman Chemical Human Arginine Vasopressin Receptor 1A Reporter Assay System, 384 Well
SDP
Catalog No. 502944987
Encompass_Preferred
INDIGO's assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to AVPR1A Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator Vasopressin, Luciferase Detection Reagents, and a cell culture-ready assay plate.
Catalog No. 50-294-4987 Supplier Cayman Chemical Supplier No. 41011384 WELL
INDIGO's assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to AVPR1A Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator Vasopressin, Luciferase Detection Reagents, and a cell culture-ready assay plate.