Learn More
Cayman Chemical Human c-MET / Hepatocyte Growth Factor Receptor Reporter Assay System, 384 Well
SDP
Catalog No. 502945859
Encompass_Preferred
This assay kit provides the convenience of an all-inclusive cell-based reporter assay system for c-MET signaling. In addition to Reporter Cells, included are an optimized culture medium for use in reviving the cryopreserved cells, a culture medium for use in preparing test sample treatments, the physiological activator HGF as a positive control, Luciferase Detection Reagents, and a cell culture-ready assay plate.
Catalog No. 50-294-5859 Supplier Cayman Chemical Supplier No. 41008384 WELL
This assay kit provides the convenience of an all-inclusive cell-based reporter assay system for c-MET signaling. In addition to Reporter Cells, included are an optimized culture medium for use in reviving the cryopreserved cells, a culture medium for use in preparing test sample treatments, the physiological activator HGF as a positive control, Luciferase Detection Reagents, and a cell culture-ready assay plate.