Learn More
Cayman Chemical Human Cannabinoid Type 1 Receptor Reporter Assay System, 3x32 Well
SDP
Catalog No. 502949654
Encompass_Preferred
CB1R Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB19001-32, IB19001, IB19002]
Catalog No. 50-294-9654 Supplier Cayman Chemical Supplier No. 342563X32 WELL
CB1R Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB19001-32, IB19001, IB19002]