Learn More
Cayman Chemical Human Constitutive Androstane Receptor (isoform 3) Reporter Assay System, 3x32 Well
SDP
Catalog No. 502972585
Encompass_Preferred
CAR3 Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, cell titer adjustments, or pre- incubation of reporter cells prior to assay setup. [INDIGO Catalog No. IB00901-32, IB00901, IB00902]
Catalog No. 50-297-2585 Supplier Cayman Chemical Supplier No. 157473X32 WELL
CAR3 Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, cell titer adjustments, or pre- incubation of reporter cells prior to assay setup. [INDIGO Catalog No. IB00901-32, IB00901, IB00902]