Learn More
Cayman Chemical Human Constitutive Androstane Receptor, isoform 2 Reporter Assay System, 3x32 Well
SDP
Catalog No. 502938831
Encompass_Preferred
CAR-2 Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB00921-32, IB00921, IB00922]
Catalog No. 50-293-8831 Supplier Cayman Chemical Supplier No. 270973X32 WELL
CAR-2 Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB00921-32, IB00921, IB00922]