Learn More
Cayman Chemical Humn EpIDrmal Growth Factr
SDP
Catalog No. 502947679
Encompass_Preferred
Reporter Cells are transiently transfected and prepared as frozen stocks using INDIGO's proprietary CryoMite™ process. This cryo-preservation method allows for the immediate dispensing of of healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup. [INDIGO Catalog Item Nos. IB13001, IB13001-32, IB13002]
Catalog No. 50-294-7679 Supplier Cayman Chemical Supplier No. 325803X32 WELL
Reporter Cells are transiently transfected and prepared as frozen stocks using INDIGO's proprietary CryoMite™ process. This cryo-preservation method allows for the immediate dispensing of of healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup. [INDIGO Catalog Item Nos. IB13001, IB13001-32, IB13002]