Learn More
Cayman Chemical Human Farnesoid X Receptor Reporter Assay System, 384 Well
SDP
Catalog No. 502972273
Encompass_Preferred
FXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB00601-32, IB00601, IB00602]
Catalog No. 50-297-2273 Supplier Cayman Chemical Supplier No. 15741384 WELL
FXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB00601-32, IB00601, IB00602]